7-Octen-1-ol ≥96.0%

Supplier: TCI America

O0280-25ML O0280-5ML
TCO0280-025MLEA 1092.92 CAD
TCO0280-025ML TCO0280-5ML
7-Octen-1-ol ≥96.0%
7-Octen-1-ol

CAS Number: 13175-44-5
MDL Number: MFCD00798076
Molecular Formula: C8H16O
Molecular Weight: 128.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 66
Specific Gravity (20/20): 0.85

Formula: C₈H₁₆O
Boiling Pt: 66 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00798076
CAS Number: 13175-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01d64af65451bd71cd260c8ffdabda08806ee374&vid=01d64af65451bd71cd260c8ffdabda08806ee374&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR