Triethyl orthodichloroacetate

Supplier: TCI America

Synonyms: 1,1,1-Triethoxy-2,2-dichloroethane, Orthodichloroacetic Acid Triethyl Ester

O0274-1G O0274-5G
TCO0274-001GEA 45.82 CAD
TCO0274-001G TCO0274-5G
Triethyl orthodichloroacetate
Triethyl orthodichloroacetate

CAS Number: 54567-92-9
MDL Number: MFCD01861293
Molecular Formula: C8H16Cl2O3
Molecular Weight: 231.11
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 195
Specific Gravity (20/20): 1.13
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,1,1-Triethoxy-2,2-dichloroethane, Orthodichloroacetic Acid Triethyl Ester

Formula: C₈H₁₆Cl₂O₃
MW: 231.11 g/mol
Boiling Pt: 195 °C
Density: 1.13 (20 °C)
MDL Number: MFCD01861293
CAS Number: 54567-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3695f48dbe7880374ffa4e5e33acb97e69a2ea90&vid=3695f48dbe7880374ffa4e5e33acb97e69a2ea90&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR