1-Oxa-2-oxo-3-thiaindolizinium chloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

O0270-5G O0270-1G
TCO0270-005GEA 333.38 CAD
TCO0270-005G TCO0270-001G
1-Oxa-2-oxo-3-thiaindolizinium chloride ≥98.0% (by total nitrogen basis)
1-Oxa-2-oxo-3-thiaindolizinium chloride

[for Source of Alkyl Radical]
CAS Number: 89025-51-4
MDL Number: MFCD00134271
Molecular Formula: C6H4ClNO2S
Molecular Weight: 189.61
Purity/Analysis Method: >98.0% (N)
Form: Crystal


Other Product Names / Synonyms: 2-Oxo[1.4.2]oxathiazolo[2,3-a]pyridinium Chloride

Formula: C₆H₄ClNO₂S
MDL Number: MFCD00134271
CAS Number: 89025-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f5b0d41a9340155adc854e390bd8915bada78bd&vid=2f5b0d41a9340155adc854e390bd8915bada78bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR