N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-18, SB-18) ≥97.0% for synthesis

Supplier: TCI America

Synonyms: Stearyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl sulfobetaine, 3-[Dimethyl(octadecyl)ammonio]propane-1-sulphonate, Stearyl sulphobetaine, 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, Octadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt, Octadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(stearyl)ammonio]propane-1-sulphonate

O0261-25G O0261-250G
TCO0261-025GEA 49.79 CAD
TCO0261-025G TCO0261-250G
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-18, SB-18) ≥97.0% for synthesis
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-18, SB-18)

CAS Number: 13177-41-8
MDL Number: MFCD00043218
Molecular Formula: C23H49NO3S
Molecular Weight: 419.71
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 260
Flash Point (°C): 110


Product Categories: Materials Chemistry, Amphoteric Surfactants
Other Product Names / Synonyms: 3-[Dimethyl(octadecyl)ammonio]propane-1-sulfonate, 3-[Dimethyl(stearyl)ammonio]propane-1-sulfonate, Stearyl Sulfobetaine, Stearyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt

Formula: C₂₃H₄₉NO₃S
Melting Pt: 260 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043218
CAS Number: 13177-41-8
EINECS: 236-124-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47423d0e031d4e7592193df6cab765ba84cbab6&vid=f47423d0e031d4e7592193df6cab765ba84cbab6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR