Octafluoroadipic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
O0260-5G
O0260-25G
TCO0260-005GEA
82.16
CAD
TCO0260-005G
TCO0260-25G
Octafluoroadipic acid ≥98.0% (by GC, titration analysis)
Octafluoroadipic acid
CAS Number: 336-08-3
MDL Number: MFCD00066405
Molecular Formula: C6H2F8O4
Molecular Weight: 290.07
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid
Formula:
C₆H₂F₈O₄ Melting Pt: 134 °C |
MDL Number:
MFCD00066405 CAS Number: 336-08-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...