3-n-Octylpyrrole ≥98.0%
Supplier: TCI America
Synonyms:
3-Octyl-1H-pyrrole
O0241-1G
O0241-5G
TCO0241-001GEA
290.88
CAD
TCO0241-001G
TCO0241-005G
3-n-Octylpyrrole ≥98.0%
3-n-Octylpyrrole
CAS Number: 118799-18-1
MDL Number: MFCD00191474
Molecular Formula: C12H21N
Molecular Weight: 179.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 0.88
Formula:
C₁₂H₂₁N Boiling Pt: 111 °C Density: 0.88 (20 °C) |
MDL Number:
MFCD00191474 CAS Number: 118799-18-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...