4,4'-Oxydiphthalic anhydride ≥98.0% (by HPLC)

Supplier: TCI America

O0237-25G O0237-100G
TCO0237-025GEA 109.18 CAD
TCO0237-025G TCO0237-100G
4,4'-Oxydiphthalic anhydride ≥98.0% (by HPLC)
4,4'-Oxydiphthalic anhydride

CAS Number: 1823-59-2
MDL Number: MFCD00039144
Molecular Formula: C16H6O7
Molecular Weight: 310.22
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 228
Flash Point (°C): 93

Formula: C₁₆H₆O₇
Melting Pt: 228 °C
Flash Pt: 93 °C
MDL Number: MFCD00039144
CAS Number: 1823-59-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR