Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl]oxalate ≥98.0%
Supplier: TCI America
Synonyms:
Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Oxalate, Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate, Oxalic Acid Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Ester, Oxalic Acid Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Ester
CAS Number: 30431-54-0
MDL Number: MFCD00191674
Molecular Formula: C26H24Cl6O8
Molecular Weight: 677.17
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 83
Product Categories: Analytical Science, Oxalates (Chemiluminescence Reagents)
Other Product Names / Synonyms: Oxalic Acid Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Ester, Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Oxalate, Oxalic Acid Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Ester
Formula:
C₂₆H₂₄Cl₆O₈ MW: 677.17 g/mol Melting Pt: 83 °C |
MDL Number:
MFCD00191674 CAS Number: 30431-54-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...