2,3,7,8,12,13,17,18-Octaethylporphyrin ≥95.0% (by total nitrogen basis)

Supplier: TCI America

O0234-1G O0234-100MG
TCO0234-001GEA 993.82 CAD
TCO0234-001G TCO0234-100MG
2,3,7,8,12,13,17,18-Octaethylporphyrin ≥95.0% (by total nitrogen basis)
2,3,7,8,12,13,17,18-Octaethylporphyrin

CAS Number: 2683-82-1
MDL Number: MFCD00009627
Molecular Formula: C36H46N4
Molecular Weight: 534.79
Purity/Analysis Method: >95.0% (N)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins
Other Product Names / Synonyms: 2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine

Formula: C₃₆H₄₆N₄
MDL Number: MFCD00009627
CAS Number: 2683-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60ca1fd4b2877e8d5397914d40c28bad491f4ff1&vid=60ca1fd4b2877e8d5397914d40c28bad491f4ff1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR