Diethyl-3-oxopimelate ≥96.0% (by titrimetric analysis)
Supplier: TCI America
O0229-1G
O0229-5G
TCO0229-1GEA
123.24
CAD
TCO0229-1G
TCO0229-005G
Diethyl-3-oxopimelate ≥96.0% (by titrimetric analysis)
Diethyl-3-oxopimelate
CAS Number: 40420-22-2
MDL Number: MFCD00009214
Molecular Formula: C11H18O5
Molecular Weight: 230.26
Purity/Analysis Method: >96.0% (T)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 113
Specific Gravity (20/20): 1.08
Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Diethyl 3-Oxoheptanedioate, 3-Oxoheptanedioic Acid Diethyl Ester, 3-Oxopimelic Acid Diethyl Ester
Formula:
C₁₁H₁₈O₅ Boiling Pt: 132 °C Density: 1.08 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00009214 CAS Number: 40420-22-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...