5-Amino-2-(4-aminophenyl)benzofuran ≥98.0%

Supplier: TCI America

Synonyms: 2-(4-Aminophenyl)benzofuran-5-amine, 2-(4-Aminophenyl)-5-benzofuranamine

A1966-1G A1966-5G
TCA1966-1GEA 346.02 CAD
TCA1966-1G TCA1966-5G
5-Amino-2-(4-aminophenyl)benzofuran ≥98.0%
5-Amino-2-(4-aminophenyl)benzofuran

CAS Number: 84102-58-9
Molecular Formula: C14H12N2O
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 198


Other Product Names / Synonyms: 2-(4-Aminophenyl)-5-benzofuranamine

Formula: C₁₄H₁₂N₂O
MW: 224.26 g/mol
Melting Pt: 198 °C
MDL Number: MFCD10000946
CAS Number: 84102-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37eeff5d027380b810a8247b9341490c04ca551b&vid=37eeff5d027380b810a8247b9341490c04ca551b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR