trans-9-Octadecenol ≥99.0%
Supplier: TCI America
Synonyms:
Elaidyl Alcohol
O0228-1G
O0228-5G
TCO0228-001GEA
453.46
CAD
TCO0228-001G
TCO0228-005G
trans-9-Octadecenol ≥99.0%
trans-9-Octadecenol
CAS Number: 506-42-3
MDL Number: MFCD00063218
Molecular Formula: C18H36O
Molecular Weight: 268.49
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 333
Melting point (°C): 37
Storage Temperature: 0-10°C
Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Elaidyl Alcohol
Formula:
C₁₈H₃₆O MW: 268.49 g/mol Boiling Pt: 333 °C Melting Pt: 37 °C |
MDL Number:
MFCD00063218 CAS Number: 506-42-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...