cis,cis-9,12-Octadecadien-1-ol ≥95.0%

Supplier: TCI America

O0225-5ML O0225-25ML
TCO0225-005MLEA 420.28 CAD
TCO0225-005ML TCO0225-25ML
cis,cis-9,12-Octadecadien-1-ol ≥95.0%
cis,cis-9,12-Octadecadien-1-ol

CAS Number: 506-43-4
MDL Number: MFCD00056667
Molecular Formula: C18H34O
Molecular Weight: 266.47
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Linoleyl Alcohol

Formula: C₁₈H₃₄O
Boiling Pt: 155 °C
Density: 0.86 (20 °C)
MDL Number: MFCD00056667
CAS Number: 506-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3670f80da1538678b86f39baacb9545307abfe6&vid=c3670f80da1538678b86f39baacb9545307abfe6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR