1-Octadecylpyrrole ≥95.0%

Supplier: TCI America

O0219-1G O0219-5G
TCO0219-001GEA 223.1 CAD
TCO0219-001G TCO0219-005G
1-Octadecylpyrrole ≥95.0%
1-Octadecylpyrrole

CAS Number: 89601-24-1
MDL Number: MFCD00144990
Molecular Formula: C22H41N
Molecular Weight: 319.58
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 39
Storage Temperature: 0-10°C

Formula: C₂₂H₄₁N
Boiling Pt: 182 °C
Melting Pt: 39 °C
MDL Number: MFCD00144990
CAS Number: 89601-24-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77d300b847812f830b2e4560719e3446663a8da9&vid=77d300b847812f830b2e4560719e3446663a8da9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR