Ethyl-3-oxoheptanoate ≥95.0%

Supplier: TCI America

O0216-10G O0216-25G
TCO0216-010GEA 274.92 CAD
TCO0216-010G TCO0216-25G
Ethyl-3-oxoheptanoate ≥95.0%
Ethyl-3-oxoheptanoate

CAS Number: 7737-62-4
MDL Number: MFCD00143188
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Bioscience, Lipids
Other Product Names / Synonyms: Ethyl 3-Oxoenanthate, 3-Oxoheptanoic Acid Ethyl Ester, 3-Ketoheptanoic Acid Ethyl Ester, 3-Oxoenanthic Acid Ethyl Ester

Formula: C₉H₁₆O₃
Boiling Pt: 111 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00143188
CAS Number: 7737-62-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ede739d02486d4aa8db50b936e7bbb44bd21aa1&vid=9ede739d02486d4aa8db50b936e7bbb44bd21aa1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR