Methyl-2-oxo-1-pyrrolidineacetate ≥97.0%

Supplier: TCI America

O0205-5G O0205-25G
TCO0205-005GEA 225.94 CAD
TCO0205-005G TCO0205-25G
Methyl-2-oxo-1-pyrrolidineacetate ≥97.0%
Methyl-2-oxo-1-pyrrolidineacetate

CAS Number: 59776-88-4
MDL Number: MFCD00003198
Molecular Formula: C7H11NO3
Molecular Weight: 157.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 2-Oxo-1-pyrrolidineacetic Acid Methyl Ester, 2-Pyrrolidinone-1-acetic Acid Methyl Ester

Formula: C₇H₁₁NO₃
Density: 1.17 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00003198
CAS Number: 59776-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67af2f3668b5bdaa503ce771f7d8cbccce9644b&vid=c67af2f3668b5bdaa503ce771f7d8cbccce9644b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR