(±)-1-Octyn-3-ol ≥98.0%

Supplier: TCI America
Warning

O0196-250ML O0196-25ML
TCO0196-250MLEA 509.08 CAD
TCO0196-250ML TCO0196-025ML
(±)-1-Octyn-3-ol ≥98.0%
(±)-1-Octyn-3-ol

CAS Number: 818-72-4
MDL Number: MFCD00004588
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 64
Specific Gravity (20/20): 0.87

Formula: C₈H₁₄O
Boiling Pt: 83 °C
Melting Pt: 0 °C
Density: 0.87 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00004588
CAS Number: 818-72-4
EINECS: 212-455-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0624dac1a06169803189f9f9b23953807281037f&vid=0624dac1a06169803189f9f9b23953807281037f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR