5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥98.0% (by HPLC)

Supplier: TCI America

A1950-25G A1950-5G
TCA1950-25GEA 36.34 CAD
TCA1950-25G TCA1950-5G
5-Amino-1,3-dihydro-2H-benzimidazol-2-one ≥98.0% (by HPLC)
5-Amino-1,3-dihydro-2H-benzimidazol-2-one

CAS Number: 95-23-8
MDL Number: MFCD00053555
Molecular Formula: C7H7N3O
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₇H₇N₃O
MDL Number: MFCD00053555
CAS Number: 95-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=984a275863487367a639b765924eacf74a544fc6&vid=984a275863487367a639b765924eacf74a544fc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR