1-Phenyl-1-octadecanone ≥98.0%

Supplier: TCI America

O0185-25G O0185-5G
TCO0185-025GEA 294.21 CAD
TCO0185-025G TCO0185-5G
1-Phenyl-1-octadecanone ≥98.0%
1-Phenyl-1-octadecanone

CAS Number: 6786-36-3
MDL Number: MFCD00010700
Molecular Formula: C24H40O
Molecular Weight: 344.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 243
Melting point (°C): 64


Other Product Names / Synonyms: Heptadecyl Phenyl Ketone, Stearophenone

Formula: C₆H₅C(O)(CH₂)₁₆CH₃
Boiling Pt: 243 °C
Melting Pt: 64 °C
MDL Number: MFCD00010700
CAS Number: 6786-36-3
EINECS: 229-850-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a88aa57bfca5e1816877b435b5645a94c8bd7ae9&vid=a88aa57bfca5e1816877b435b5645a94c8bd7ae9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR