Oxamic acid potassium salt ≥98.0%
Supplier: TCI America
Synonyms:
Potassium Oxamate, Oxamidic Acid Potassium Salt
O0169-25G
O0169-500G
TCO0169-025GEA
91.64
CAD
TCO0169-025G
TCO0169-500G
Oxamic acid potassium salt ≥98.0%
Oxamic acid potassium salt
CAS Number: 21141-31-1
MDL Number: MFCD00058475
Molecular Formula: C2H3NO3
Molecular Weight: 127.14
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Other Product Names / Synonyms: Oxamidic Acid Potassium Salt, Potassium Oxamate
Formula:
C₂H₃NO₃ |
MDL Number:
MFCD00058475 CAS Number: 21141-31-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...