5-(Acetoacetamido)-2-benzimidazolinone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 5-Acetoacetlamino benzimdazolone

A1949-250G A1949-25G
TCA1949-250GEA 396.58 CAD
TCA1949-250G TCA1949-25G
5-(Acetoacetamido)-2-benzimidazolinone ≥98.0% (by HPLC, total nitrogen)
5-(Acetoacetamido)-2-benzimidazolinone

CAS Number: 26576-46-5
MDL Number: MFCD00444508
Molecular Formula: C11H11N3O3
Molecular Weight: 233.23
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₁₁H₉N₃O₃
MW: 231.21 g/mol
MDL Number: MFCD00444508
CAS Number: 26576-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f7469c1c25d245287db3523023f5b03ba3d6b20&vid=6f7469c1c25d245287db3523023f5b03ba3d6b20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR