4-(2-Octylamino)diphenylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Phenyl-N'-(2-octyl)-1,4-phenylenediamine, N-(1-Methylheptyl)-N'-phenyl-1,4-phenylenediamine

O0166-25G O0166-500G
TCO0166-025GEA 26.86 CAD
TCO0166-025G TCO0166-500G
4-(2-Octylamino)diphenylamine ≥98.0% (by GC, titration analysis)
4-(2-Octylamino)diphenylamine

CAS Number: 15233-47-3
MDL Number: MFCD00059441
Molecular Formula: C20H28N2
Molecular Weight: 296.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: N-(1-Methylheptyl)-N'-phenyl-1,4-phenylenediamine, N-Phenyl-N'-(2-octyl)-1,4-phenylenediamine

Formula: C₂₀H₂₈N₂
MW: 296.46 g/mol
Density: 1.00 (20 °C)
MDL Number: MFCD00059441
CAS Number: 15233-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f90701a4d12a1432d4a1c3a1540f69d0efdbafc&vid=5f90701a4d12a1432d4a1c3a1540f69d0efdbafc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR