(±)-3-Octanol ≥98.0%

Supplier: TCI America
Warning

O0121-25ML O0121-500ML O0121-100ML
TCO0121-025MLEA 66.68 CAD
TCO0121-025ML TCO0121-500ML TCO0121-100ML
(±)-3-Octanol ≥98.0%
(±)-3-Octanol

CAS Number: 589-98-0
MDL Number: MFCD00004590
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 177
Flash Point (°C): 68
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 3-Octyl Alcohol

Formula: C₈H₁₈O
Boiling Pt: 177 °C
Melting Pt: –45 °C
Density: 0.82 (20 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00004590
CAS Number: 589-98-0
EINECS: 209-667-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53306b86c8ffe59cb4402885bc181460869a35b0&vid=da1db8c476292949544e34b4ca69307f2dd0459f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR