Diphenyl oxalate ≥98.0%

Supplier: TCI America

O0111-25G O0111-500G O0111-100G
TCO0111-025GEA 238.9 CAD
TCO0111-025G TCO0111-500G TCO0111-100G
Diphenyl oxalate ≥98.0%
Diphenyl oxalate

CAS Number: 3155-16-6
MDL Number: MFCD00059682
Molecular Formula: C14H10O4
Molecular Weight: 242.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: Oxalic Acid Diphenyl Ester

Formula: C₁₄H₁₀O₄
Melting Pt: 136 °C
MDL Number: MFCD00059682
CAS Number: 3155-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54de219c6dde3b5889ad3bb8c2a9a1a9198bd9af&vid=54de219c6dde3b5889ad3bb8c2a9a1a9198bd9af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR