2,2'-Oxydiethanethiol ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Bis(2-mercaptoethyl) ether, 2-Mercaptoethyl ether, 2,2'-Oxydiethanethiol, Di(2-mercaptoethyl) ether

O0106-25ML O0106-500ML
TCO0106-025MLEA 208.62 CAD
TCO0106-025ML TCO0106-500ML
2,2'-Oxydiethanethiol ≥95.0% (by GC, titration analysis)
2,2'-Oxydiethanethiol

CAS Number: 2150-02-9
MDL Number: MFCD00004889
Molecular Formula: C4H10OS2
Molecular Weight: 138.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 90
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Di(2-mercaptoethyl) Ether, 2-Mercaptoethyl Ether, 2,2'-Oxydiethanethiol

Formula: C₄H₁₀OS₂
MW: 138.25 g/mol
Boiling Pt: 217 °C
Density: 1.12 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00004889
CAS Number: 2150-02-9
EINECS: 218-421-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fad02a383bbf145b3c5c1228e993f760f7019f7&vid=8fad02a383bbf145b3c5c1228e993f760f7019f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR