Solvent Black 5

Supplier: TCI America

Synonyms: 2-Amino-N,N-dipropylacetamide hydrochlorideNigrosin soluble in spiritNigrosin B, alcohol soluble

O0097-25G
TCO0097-025GEA 37.6 CAD
TCO0097-025G
Solvent Black 5
Solvent black 5

CAS Number: 11099-03-9
MDL Number: MFCD00071316
Form: Crystal
Color: Black


Product Categories: Bioscience, Stains & Dyes

Formula: C₈H₁₉ClN₂O
MDL Number: MFCD00071316
CAS Number: 11099-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR