Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)

Supplier: TCI America

O0096-5G O0096-25G
TCO0096-005GEA 57.83 CAD
TCO0096-005G TCO0096-25G
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan Red 7B (Fat Red Bluish, Solvent Red 19, Fat Red 7B)

CAS Number: 6368-72-5
MDL Number: MFCD00003904
Molecular Formula: C24H21N5
Molecular Weight: 379.47
Form: Crystal
Color: Yellow Red
Melting point (°C): 140
Lambda max.: 534 nm (MeOH)


Other Product Names / Synonyms: Fat Red 7B, Solvent Red 19, Sudan Red 7B

Formula: C₂₄H₂₁N₅
Melting Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00003904
CAS Number: 6368-72-5
EINECS: 228-862-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=885b4f06dc02af334a8138b58a27a4b2de8fba98&vid=885b4f06dc02af334a8138b58a27a4b2de8fba98&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR