5-Amino-4,6-dichloro-2-methylpyrimidine ≥98.0%

Supplier: TCI America

Synonyms: 4,6-dichloro-2-methylpyrimidin-5-amine

A1946-25G A1946-5G
TCA1946-25GEA 761.56 CAD
TCA1946-25G TCA1946-5G
5-Amino-4,6-dichloro-2-methylpyrimidine ≥98.0%
5-Amino-4,6-dichloro-2-methylpyrimidine

CAS Number: 39906-04-2
MDL Number: MFCD00194053
Molecular Formula: C5H5Cl2N3
Molecular Weight: 178.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73

Formula: C₅H₅Cl₂N₃
MW: 178.0200 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00194053
CAS Number: 39906-04-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=143e633880ab325ebc69251515bb9cc2fcbafdcb&vid=143e633880ab325ebc69251515bb9cc2fcbafdcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR