Dioctyl ether ≥96.0%

Supplier: TCI America

O0047-25ML O0047-500ML O0047-100ML
TCO0047-025MLEA 72.68 CAD
TCO0047-025ML TCO0047-500ML TCO0047-100ML
Dioctyl ether ≥96.0%
Dioctyl ether

CAS Number: 629-82-3
MDL Number: MFCD00009563
Molecular Formula: C16H34O
Molecular Weight: 242.45
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 287
Melting point (°C): -7
Flash Point (°C): 100
Specific Gravity (20/20): 0.81


Other Product Names / Synonyms: Di-n-octyl Ether

Formula: C₁₆H₃₄O
Boiling Pt: 287 °C
Melting Pt: –7 °C
Density: 0.81 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009563
CAS Number: 629-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eee7f8d98c5efd13473658c842973c4ea55f623f&vid=eee7f8d98c5efd13473658c842973c4ea55f623f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR