1-Acetyl-2-imidazolidinone ≥98.0%

Supplier: TCI America

Synonyms: 1-Acetylimidazolidin-2-one

A1945-25G A1945-5G
TCA1945-25GEA 335.59 CAD
TCA1945-25G TCA1945-5G
1-Acetyl-2-imidazolidinone ≥98.0%
1-Acetyl-2-imidazolidinone

CAS Number: 5391-39-9
MDL Number: MFCD00988254
Molecular Formula: C5H8N2O2
Molecular Weight: 128.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 185

Formula: C₅H₈N₂O₂
Melting Pt: 185 °C
MDL Number: MFCD00988254
CAS Number: 5391-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0913015bf87f47c26a84778adc4b8ac10525f72b&vid=0913015bf87f47c26a84778adc4b8ac10525f72b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR