2-Octenylsuccinic anhydride (cis and trans mixture) ≥95.0%
Supplier: TCI America
Synonyms:
3-(Oct-2-en-1-yl)dihydrofuran-2,5-dione
O0040-500G
O0040-25G
TCO0040-500GEA
420.28
CAD
TCO0040-500G
TCO0040-025G
2-Octenylsuccinic anhydride (cis and trans mixture) ≥95.0%
2-Octenylsuccinic anhydride (cis and trans mixture)
CAS Number: 42482-06-4
MDL Number: MFCD00047133
Molecular Formula: C12H18O3
Molecular Weight: 210.27
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 147
Specific Gravity (20/20): 1.03
Formula:
C₁₂H₁₈O₃ Density: 1.03 (20 °C) Flash Pt: 147 °C |
MDL Number:
MFCD00047133 CAS Number: 42482-06-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...