Amyl-n-octanoate ≥98.0%
Supplier: TCI America
O0028-25ML
O0028-500ML
TCO0028-025MLEA
47.4
CAD
TCO0028-025ML
TCO0028-500ML
Amyl-n-octanoate ≥98.0%
Amyl-n-octanoate
CAS Number: 638-25-5
MDL Number: MFCD00048919
Molecular Formula: C13H26O2
Molecular Weight: 214.35
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Specific Gravity (20/20): 0.86
Other Product Names / Synonyms: n-Octanoic Acid Amyl Ester, Pentyl n-Octanoate
Formula:
C₁₃H₂₆O₂ Boiling Pt: 260 °C Density: 0.86 (20 °C) |
MDL Number:
MFCD00048919 CAS Number: 638-25-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...