Aniline-2,5-disulfonic acid monosodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Sodium Hydrogen 2-Amino-1,4-benzenedisulphonate, 2,5-Disulfoaniline Monosodium Salt, Aniline-2,5-disulphonic Acid Monosodium Salt, Aniline-2,5-disulfonic Acid Monosodium Salt, Sodium 2-amino-4-sulfobenzenesulfonate, 2,5-Disulphoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate, 2-Amino-1,4-benzenedisulfonic acid Monosodium Salt, Sodium 2-amino-4-sulphobenzenesulphonate, 2-Amino-1,4-benzenedisulphonic acid Monosodium Salt

A0263-500G A0263-25G
TCA0263-500GEA 478.74 CAD
TCA0263-500G TCA0263-025G
Aniline-2,5-disulfonic acid monosodium salt ≥98.0% (by HPLC, titration analysis)
Aniline-2,5-disulfonic acid monosodium salt

CAS Number: 24605-36-5
MDL Number: MFCD00078393
Molecular Formula: C6H7NO6S2
Molecular Weight: 275.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Amino-1,4-benzenedisulfonic Acid Monosodium Salt, 2,5-Disulfoaniline Monosodium Salt, Sodium Hydrogen 2-Amino-1,4-benzenedisulfonate

Formula: C₆H₇NO₆S₂
MW: 275.22 g/mol
MDL Number: MFCD00078393
CAS Number: 24605-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fcc64de91fcb7a20f84cd2073da78b25ea3ff54&vid=8fcc64de91fcb7a20f84cd2073da78b25ea3ff54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR