Octadecenylsuccinic anhydride (mixture of isomers) ≥93.0%

Supplier: TCI America

O0013-25G O0013-500G
TCO0013-025GEA 56.88 CAD
TCO0013-025G TCO0013-500G
Octadecenylsuccinic anhydride (mixture of isomers) ≥93.0%
Octadecenylsuccinic anhydride (mixture of isomers)

CAS Number: 28777-98-2
MDL Number: MFCD00014545
Molecular Formula: C22H38O3
Molecular Weight: 350.54
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 70
Flash Point (°C): 205

Formula: C₂₂H₃₈O₃
Melting Pt: 70 °C
Flash Pt: 205 °C
MDL Number: MFCD00014545
CAS Number: 28777-98-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4441ec40176d7bf2465a5364be4141eef0010d2c&vid=4441ec40176d7bf2465a5364be4141eef0010d2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR