Phenyl stearate ≥98.0%

Supplier: TCI America

Synonyms: Stearic Acid Phenyl Ester

O0007-25G O0007-5G
TCO0007-025GEA 821.6 CAD
TCO0007-025G TCO0007-5G
Phenyl stearate ≥98.0%
Phenyl stearate

CAS Number: 637-55-8
MDL Number: MFCD00026663
Molecular Formula: C24H40O2
Molecular Weight: 360.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 53


Other Product Names / Synonyms: Stearic Acid Phenyl Ester

Formula: C₂₄H₄₀O₂
MW: 360.58 g/mol
Boiling Pt: 267 °C
Melting Pt: 53 °C
MDL Number: MFCD00026663
CAS Number: 637-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62178a1556aa30faf37033a13b427ed0478e76c9&vid=62178a1556aa30faf37033a13b427ed0478e76c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR