2-Naphthyl trifluoromethanesulfonate ≥95.0%

Supplier: TCI America

Synonyms: 2-Naphthyl Trifluoromethanesulphonate2-Naphthyl TriflateTrifluoromethanesulfonic acid 2-Naphthyl Ester2-Naphthyl TrifluoromethanesulfonateTrifluoromethanesulphonic acid 2-Naphthyl Ester

N0845-5G N0845-1G
TCN0845-1GEA 121.25 CAD
TCN0845-1G TCN0845-5G
2-Naphthyl trifluoromethanesulfonate ≥95.0%
2-Naphthyl trifluoromethanesulfonate

CAS Number: 3857-83-8
MDL Number: MFCD00192341
Molecular Formula: C11H7F3O3S
Molecular Weight: 276.23
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 87
Melting point (°C): 32
Flash Point (°C): 113


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Trifluoromethanesulfonic Acid 2-Naphthyl Ester, 2-Naphthyl Triflate

Formula: C₁₁H₇F₃O₃S
MW: 276.23 g/mol
Boiling Pt: 87 °C
Melting Pt: 32 °C
Flash Pt: 113 °C
MDL Number: MFCD00192341
CAS Number: 3857-83-8
https://ca.vwr.com/stibo/web/std.lang.all/00/40/28990040.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR