Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

N0844-5G N0844-25G
TCN0844-5GEA 110.6 CAD
TCN0844-5G TCN0844-25G
Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)
Nortropinone hydrochloride

CAS Number: 25602-68-0
MDL Number: MFCD03613582
Molecular Formula: C7H11NO
Molecular Weight: 161.63
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 204


Product Categories: Bioscience, Tropane Alkaloids
Other Product Names / Synonyms: 3-Nortropanone Hydrochloride, 8-Azabicyclo[3.2.1]octan-3-one Hydrochloride

Formula: C₇H₁₂ClNO
MW: 161.63 g/mol
Melting Pt: 204 °C
MDL Number: MFCD03613582
CAS Number: 25602-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0f89a7e6af95ce3e1d292af1be5bb4fb870206c&vid=d0f89a7e6af95ce3e1d292af1be5bb4fb870206c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR