Nicorandil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-[2-(Nitrooxy)ethyl]pyridine-3-carboxamide, 2-(Pyridine-3-carboxamido)ethyl Nitrate, 2-(Pyridine-3-carbonylamino)ethyl nitrate

N0837-1G N0837-5G N0837-100MG
TCN0837-1GEA 185.18 CAD
TCN0837-1G TCN0837-5G TCN0837-100MG
Nicorandil ≥98.0% (by HPLC, titration analysis)
Nicorandil

CAS Number: 65141-46-0
MDL Number: MFCD00186520
Molecular Formula: C8H9N3O4
Molecular Weight: 211.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 90


Product Categories: Bioscience, Other Pharmaceutical Ingredients, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 2-(Pyridine-3-carboxamido)ethyl Nitrate, N-[2-(Nitrooxy)ethyl]pyridine-3-carboxamide

Formula: C₈H₉N₃O₄
MW: 211.18 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00186520
CAS Number: 65141-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f2535b429068e6893b2e1481144e92e417a9ced&vid=0f2535b429068e6893b2e1481144e92e417a9ced&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR