2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol ≥97.0%

Supplier: TCI America

Synonyms: 1H,1H-Nonafluoro-1-pentanol, 1H,1H-Perfluoropentan-1-ol

N0810-1G N0810-25G N0810-5G
TCN0810-1GEA 55.3 CAD
TCN0810-1G 76220-356 76220-358
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol ≥97.0%
2,2,3,3,4,4,5,5,5-Nonafluoro-1-pentanol

CAS Number: 355-28-2
MDL Number: MFCD00236714
Molecular Formula: C5H3F9O
Molecular Weight: 250.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Specific Gravity (20/20): 1.66


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Perfluoro-1-pentanol, (Perfluorobutyl)methanol

Formula: C₅H₃F₉O
MW: 250.06 g/mol
Boiling Pt: 111 °C
Density: 1.66 (20 °C)
MDL Number: MFCD00236714
CAS Number: 355-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b69d0e58b1264e5d5768552cf9d52162774b29&vid=e2b69d0e58b1264e5d5768552cf9d52162774b29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR