2-Nitro-p-cymene ≥95.0%

Supplier: TCI America

N0801-25G N0801-5G
TCN0801-25GEA 876.9 CAD
TCN0801-25G TCN0801-5G
2-Nitro-p-cymene ≥95.0%
2-Nitro-p-cymene

CAS Number: 943-15-7
MDL Number: MFCD00007176
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 113
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene

Formula: C₁₀H₁₃NO₂
Boiling Pt: 134 °C
Density: 1.07 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00007176
CAS Number: 943-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5af18346327e376ab1730cb84267a75f47a4f14a&vid=5af18346327e376ab1730cb84267a75f47a4f14a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR