3-Nitrothioanisole ≥98.0%
Supplier: TCI America
N0800-5G
N0800-25G
TCN0800-5GEA
87.53
CAD
TCN0800-5G
TCN0800-25G
3-Nitrothioanisole ≥98.0%
3-Nitrothioanisole
CAS Number: 2524-76-7
MDL Number: MFCD00024501
Molecular Formula: C7H7NO2S
Molecular Weight: 169.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Melting point (°C): 14
Specific Gravity (20/20): 1.29
Other Product Names / Synonyms: 1-(Methylthio)-3-nitrobenzene
Formula:
C₇H₇NO₂S Boiling Pt: 92 °C Melting Pt: 14 °C Density: 1.29 (20 °C) |
MDL Number:
MFCD00024501 CAS Number: 2524-76-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...