4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
4-(1-Naphthyl)benzeneboronic acid
N0798-5G
N0798-1G
TCN0798-5GEA
120.08
CAD
TCN0798-5G
TCN0798-1G
4-(1-Naphthyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Naphthyl)phenylboronic acid
CAS Number: 870774-25-7
Molecular Formula: C16H13BO2
Molecular Weight: 248.09
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-(1-Naphthyl)benzeneboronic Acid
Formula:
C₁₆H₁₃BO₂ MW: 248.09 g/mol |
MDL Number:
MFCD08669639 CAS Number: 870774-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...