Neu5Ac-ɑ(2-3)Gal-β-MP Glycoside ≥95.0% (by HPLC)

Supplier: TCI America

N0791-10MG N0791-50MG
TCN0791-10MGEA 451.25 CAD
TCN0791-10MG TCN0791-50MG
Neu5Ac-ɑ(2-3)Gal-β-MP Glycoside ≥95.0% (by HPLC)
Neu5Ac-alpha(2-3)Gal-beta-MP Glycoside

CAS Number: 159922-54-0
Molecular Formula: C24H35NO15
Molecular Weight: 577.54
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Disaccharides, Sialylated Oligosaccharides, Glycosides, p-Methoxyphenyl Glycosides

Formula: C₂₄H₃₅NO₁₅
CAS Number: 159922-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88b0b3c462e307bbb2141b97e929de95090a6963&vid=88b0b3c462e307bbb2141b97e929de95090a6963&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR