Neopentyl glycol dimethacrylate ≥90.0% stabilised

Supplier: TCI America

Synonyms: 2,2-Dimethylpropane-1,3-diyl bis(2-methylacrylate), 2,2-Dimethyl-1,3-propanediol Dimethacrylate, 1,3-Bis(methacryloyloxy)-2,2-dimethylpropane

N0780-25G N0780-500G
TCN0780-25GEA 44.98 CAD
TCN0780-25G TCN0780-500G
Neopentyl glycol dimethacrylate ≥90.0% stabilised
Neopentyl glycol dimethacrylate

(stabilized with MEHQ)
CAS Number: 1985-51-9
Molecular Formula: C13H20O4
Molecular Weight: 240.30
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 140
Specific Gravity (20/20): 1.00


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Bis(methacryloyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Dimethacrylate

Formula: C₁₃H₂₀O₄
MW: 240.30 g/mol
Density: 1.00 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00048120
CAS Number: 1985-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3414fe6124be435905ce6b7fa97337c568cab4c6&vid=3414fe6124be435905ce6b7fa97337c568cab4c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR