2-Amino-3,5-dichloro-4-methylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1932-5G A1932-1G
TCA1932-5GEA 88.48 CAD
TCA1932-5G TCA1932-1G
2-Amino-3,5-dichloro-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-3,5-dichloro-4-methylpyridine

CAS Number: 31430-47-4
MDL Number: MFCD00129031
Molecular Formula: C6H6Cl2N2
Molecular Weight: 177.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 2-Amino-3,5-dichloro-4-picoline

Formula: C₆H₆Cl₂N₂
Melting Pt: 125 °C
MDL Number: MFCD00129031
CAS Number: 31430-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e15291792826f6ff08ee8b238a70110fd12d51f2&vid=e15291792826f6ff08ee8b238a70110fd12d51f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR