exo-Norborneol ≥98.0%

Supplier: TCI America

N0775-25G N0775-5G
TCN0775-25GEA 517.29 CAD
TCN0775-25G TCN0775-5G
exo-Norborneol ≥98.0%
exo-Norborneol

CAS Number: 497-37-0
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 128

Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 177 °C
Melting Pt: 128 °C
CAS Number: 497-37-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47c9911703de81fc3ed02580c5fdc5001310bc5f&vid=47c9911703de81fc3ed02580c5fdc5001310bc5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR