3-(1-Naphthyl)-L-alanine hydrochloride ≥97.0%
Supplier: TCI America
Synonyms:
(S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
N0773-5G
N0773-1G
TCN0773-5GEA
715.74
CAD
TCN0773-5G
TCN0773-1G
3-(1-Naphthyl)-L-alanine hydrochloride ≥97.0%
3-(1-Naphthyl)-L-alanine hydrochloride
CAS Number: 122745-10-2
Molecular Formula: C13H13NO2
Molecular Weight: 251.71
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -8.4 deg (C=1, MeOH)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride
Formula:
C₁₃H₁₃NO₂ MW: 251.71 g/mol |
CAS Number:
122745-10-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...