3-(1-Naphthyl)-D-alanine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (R)-2-Amino-3-(naphthalen-1-yl)propanoic acid hydrochloride, (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride

N0772-1G N0772-5G
TCN0772-1GEA 64.78 CAD
TCN0772-1G TCN0772-5G
3-(1-Naphthyl)-D-alanine hydrochloride ≥98.0% (by titrimetric analysis)
3-(1-Naphthyl)-D-alanine hydrochloride

CAS Number: 122745-09-9
Molecular Formula: C13H13NO2
Molecular Weight: 251.71
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 8 deg (C=1, MeOH)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-alpha-Amino-1-naphthalenepropionic Acid Hydrochloride

Formula: C₁₃H₁₃NO₂
MW: 251.71 g/mol
MDL Number: MFCD03095597
CAS Number: 122745-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0af96f330abca0dabcd0003f714d46328bb10fa&vid=b0af96f330abca0dabcd0003f714d46328bb10fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR