1,4,5,8-Naphthalenetetracarboxylic acid 60.0% (by NMR) (contains Monoanhydride)
Supplier: TCI America
N0770-25G
N0770-250G
TCN0770-25GEA
172.22
CAD
TCN0770-25G
TCN0770-250G
1,4,5,8-Naphthalenetetracarboxylic acid 60.0% (by NMR) (contains Monoanhydride)
1,4,5,8-Naphthalenetetracarboxylic acid
CAS Number: 128-97-2
Molecular Formula: C14H8O8
Molecular Weight: 304.21
Purity/Analysis Method: >60.0% (NMR)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₁₄H₈O₈ |
MDL Number:
MFCD00004012 CAS Number: 128-97-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...