5-Norbornene-2-exo,3-exo-dimethanol ≥97.0%

Supplier: TCI America

N0769-5G N0769-1G
TCN0769-5GEA 663.6 CAD
TCN0769-5G TCN0769-1G
5-Norbornene-2-exo,3-exo-dimethanol ≥97.0%
5-Norbornene-2-exo,3-exo-dimethanol

CAS Number: 699-95-6
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 97
Melting point (°C): 53
Flash Point (°C): 113


Other Product Names / Synonyms: Bicyclo[2.2.1]hept-5-ene-2-exo,3-exo-dimethanol, 5-exo,6-exo-Bis(hydroxymethyl)-2-norbornene

Formula: C₉H₁₄O₂
Boiling Pt: 97 °C
Melting Pt: 53 °C
Flash Pt: 113 °C
MDL Number: MFCD00799568
CAS Number: 699-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=638d0efd0d1eb5ab381c8075c993d048954431c9&vid=638d0efd0d1eb5ab381c8075c993d048954431c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR